Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinculin, Mouse anti-Human, Clone: VCL/2575, Novus Biologicals™

Mouse Monoclonal Antibody

$156.50 - $345.50

Specifications

Antigen Vinculin
Clone VCL/2575
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
Host Species Mouse
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB010633
View Documents
Novus Biologicals
NBP279808100UG
100 ug
Each for $345.50
Add to cart
 
NB010634
View Documents
Novus Biologicals
NBP27980820UG
20 ug
Each for $156.50
Add to cart
 
Description

Description

Vinculin Monoclonal antibody specifically detects Vinculin in Human Samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array.
Specifications

Specifications

Vinculin
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
Human
Primary
VCL/2575
Mouse
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit