Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vinculin, Mouse anti-Human, DyLight 350, Clone: VCL/2575, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP279942UV
Description
Vinculin Monoclonal antibody specifically detects Vinculin in Human Samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
Vinculin | |
VCL/2575 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Purified | |
VCL | |
Recombinant fragment (around aa 174-322) of human Vinculin protein (exact sequence is proprietary) | |
Protein A or G purified | |
RUO | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
DyLight 350 | |
50mM Sodium Borate | |
CMD1W, CMH15, Metavinculin, MVCL, vinculin | |
Mouse | |
IgG2b Kappa | |
0.1 mL | |
Cell Biology, Cellular Markers, Phospho Specific | |
Primary | |
7414 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title