Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinculin, Mouse anti-Human,Mouse, Janelia Fluor 646, Clone: VCL/2573, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP279941JF646

Catalog No. NB013065


Add to cart

Description

Description

Vinculin Monoclonal antibody specifically detects Vinculin in Human, Mouse Samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

Vinculin
VCL/2573
Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
VCL
Recombinant fragment (around aa 174-322) of human Vinculin protein (exact sequence is proprietary)
Protein A or G purified
RUO
Store at 4°C in the dark.
Monoclonal
Human, Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin)
Janelia Fluor 646
50mM Sodium Borate
CMD1W, CMH15, Metavinculin, MVCL, vinculin
Mouse
IgG1 Kappa
0.1 mL
Cell Biology, Cellular Markers, Phospho Specific
Primary
7414
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit