Learn More
Vinculin Mouse anti-Human, Mouse, Rat, Clone: 7F9, eBioscience™ Invitrogen™
Mouse Monoclonal Antibody
Manufacturer: LIFE TECHNOLOGIES 14977795

Description
The MAb 7F9 (VIIF9) recognizes human, mouse, rat, and avian vinculin and its alternatively spliced isoform, metavinculin. Vinculin is a cytoskeletal protein associated with cell-cell and cell-matrix junctions. Vinculin is involved in the anchoring of F-actin to the membrane and the regulation of E-cadherin expression. Vinculin binds to talin, paxillin, and alpha-actinin. Disregulation of vinculin alters cell adhesion, migration, and growth, which promotes cancer invasion and metastasis.
Vinculin Monoclonal antibody specifically detects Vinculin in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western BlotSpecifications
Vinculin | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P18206, P85972, Q64727 | |
Mouse | |
10 mg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot | |
7F9 | |
Unconjugated | |
VCL | |
9430097D22; AA571387; AI462105; AW545629; CMD1W; CMH15; epididymis luminal protein 114; HEL114; Metavinculin; metavinculin {ECO:0000250; MV; MVCL; RP11-178G16.3; UniProtKB:P18206}; VCL; vcl {ECO:0000250; VIN; vinc; VINC1; Vinculin; vinculin {ECO:0000250 | |
Affinity chromatography | |
RUO | |
22330, 305679, 7414 | |
Antibody | |
IgG1 κ |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.