Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinorelbine ditartrate, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
125317-39-7
C53H66N4O20
1079.119
CILBMBUYJCWATM-HJBYVSOESA-N
vinorelbine ditartrate
124080885
CCC1=CC2CC(C3=C(CN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC.C(C(C(=O)O)O)(C(=O)O)O.C(C(C(=O)O)O)(C(=O)O)O
This item is not returnable. View return policy
125317-39-7
C53H66N4O20
1079.119
CILBMBUYJCWATM-HJBYVSOESA-N
vinorelbine ditartrate
124080885
CCC1=CC2CC(C3=C(CN(C2)C1)C4=CC=CC=C4N3)(C5=C(C=C6C(=C5)C78CCN9C7C(C=CC9)(C(C(C8N6C)(C(=O)OC)O)OC(=O)C)CC)OC)C(=O)OC.C(C(C(=O)O)O)(C(=O)O)O.C(C(C(=O)O)O)(C(=O)O)O

Selective mitotic microtubule antagonist

Selective mitotic microtubule antagonist that exhibits > 20 fold selectivity over axonal microtubules. Inhibits proliferation of multiple human tumor cell lines (IC50 = 1.25 nM in HeLa cells) and blocks metaphase/anaphase transition by suppression of microtubule dynamics (IC50 = 3.8 nM). Reduces spindle length by 29% and inhibits microtubule polymerization at micromolar concentrations.
Quantity 10mg
Formula Weight 1079.1
Percent Purity >98%
Chemical Name or Material Vinorelbine ditartrate

Recommended Storage : Store at -20°C