Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vinorelbine ditartrate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective mitotic microtubule antagonist that exhibits > 20 fold selectivity over axonal microtubules. Inhibits proliferation of multiple human tumor cell lines (IC50 = 1.25 nM in HeLa cells) and blocks metaphase/anaphase transition by suppression of microtubule dynamics (IC50 = 3.8 nM). Reduces spindle length by 29% and inhibits microtubule polymerization at micromolar concentrations.
Specifications
Quantity | 10 mg |
Formula Weight | 1079.1 |
Percent Purity | >98% |
Chemical Name or Material | Vinorelbine ditartrate |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction