Vinyl acetate, 99%, stab. with 8-12ppm hydroquinone

Manufacturer: Alfa Aesar A16247AP

Quantity 500mL

 View more versions of this product

Catalog No. AAA16247AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 108-05-4
    Molecular Formula CH2CHO2CCH3
    Linear Formula CH2=CHO2CCH3
    Formula Weight 86.09
    MDL Number MFCD00008713
    UN Number UN1301
    Beilstein 1209327
    Merck Index 14,9992
    Chemical Name or Material Vinyl Acetate, stabilized with 8 to 12ppm hydroquinone

    Specifications

    Formula Weight 86.09
    Quantity 500mL
    TSCA TSCA
    Linear Formula CH2=CHO2CCH3
    Merck Index 14,9992
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and re
    Recommended Storage 2° to 8°C
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: VINYL ACETATE, STABILIZED
    RTECS Number AK0875000
    MDL Number MFCD00008713
    UN Number UN1301
    Beilstein 1209327
    Health Hazard 2 GHS H Statement
    H225
    Highly flammable liquid and vapour.
    Molecular Formula CH2CHO2CCH3
    CAS 108-05-4
    Chemical Name or Material Vinyl Acetate, stabilized with 8 to 12ppm hydroquinone
    Assay 99%

    Description

    Applications
    Vinyl acetate is used in the preparation of polyvinyl acetate emulsions and polyvinyl alcohol. It acts as a precursor to polyvinyl acetate. It serves as an ingredient in furniture-glue. It is also used to prepare copolymers with other monomers such as ethylene, acrylic acid. Further, it is used to make adhesives, paints and plastics.

    Solubility
    Slightly miscible with water.

    Notes
    Light sensitive. Incompatible with acids, bases, oxidizing agents, peroxides, chlorosulfonic acid, ethylene imine, hydrochloric acid, oleum, nitric acid, sulfuric acid and 2-aminoethanol.

    GHS H Statement
    H225
    Highly flammable liquid and vapour.

    2° to 8°C

    GHS P Statement
    P210-P280-P240-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and re

    Danger

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit