Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl acetate, MilliporeSigma™ Supelco™

Vinyl acetate is an important vinyl ester, also known as vinyl acetate monomer. It is mainly used in the production of polymers and copolymers, for coating paints, binders, textile, and paper processing industries.

Manufacturer:  MilliporeSigma Supelco 4S8486

 View more versions of this product

Catalog No. 11-102-0632


Add to cart

Description

Description

Environmental and Petroleum
Specifications

Specifications

108-05-4
MFCD00008713
86.09
Analytical Standard
88 mmHg (20°C)
0.934 g/mL (at 25°C)
72°C to 73°C
-7°C
CH3 CO2 CH=CH2
Acetoxyethylene
Neat
801°F
3 (vs. air)
-93°C
100 mg
Organic Pollutant Standards
Safety and Handling

Safety and Handling

RTECSNumber : AK0875000

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit