Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl acetate, MilliporeSigma™ Supelco™

Vinyl acetate is an important vinyl ester, also known as vinyl acetate monomer. It is mainly used in the production of polymers and copolymers, for coating paints, binders, textile, and paper processing industries.

$23.55

Specifications

Density 0.934 g/mL (at 25°C)
Grade Analytical Standard
Linear Formula CH3 CO2 CH=CH2
Physical Form Neat
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
11-102-0632
View Documents
MilliporeSigma Supelco
4S8486
100 mg
Each for $23.55
Add to cart
 
Description

Description

Environmental and Petroleum
Specifications

Specifications

0.934 g/mL (at 25°C)
Analytical Standard
CH3 CO2 CH=CH2
Neat
Safety and Handling

Safety and Handling

RTECSNumber : AK0875000

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit