Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl acetate, 99+%, stabilized, Thermo Scientific Chemicals

Catalog Number p-4510015
Click to view available options
Quantity:
25 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
108-05-4, 123-31-9
C4H6O2
86.09
XTXRWKRVRITETP-UHFFFAOYSA-N
vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
7904
CHEBI:46916
ethenyl acetate
CC(=O)OC=C
108-05-4, 123-31-9
C4H6O2
86.09
XTXRWKRVRITETP-UHFFFAOYSA-N
vinyl acetate, acetic acid ethenyl ester, acetic acid vinyl ester, ethenyl ethanoate, 1-acetoxyethylene, vinyl ethanoate, acetoxyethene, vinylacetat, vinyl acetate monomer, vinyl a monomer
7904
CHEBI:46916
ethenyl acetate
CC(=O)OC=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -93°C
Color Colorless
Density 0.9300g/mL
Boiling Point 72°C to 73°C
Flash Point -8°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Refractive Index 1.3940 to 1.396
Linear Formula CH3COOCH=CH2
Quantity 25 mL
Beilstein 02,I,63
Fieser 01,1271; 12,565
Specific Gravity 0.93
Merck Index 15,10189
Solubility Information Solubility in water: 23g/L (20°C). Other solubilities: miscible with ethanol and ether,soluble in acetone,benzene and chloroform
Stabilizer 3 to 15ppm (hydroquinone)
Viscosity 0.43 mPa.s (20°C)
Formula Weight 86.09
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Vinyl acetate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
Harmful if inhaled.
Precautionary Statement GHS P Statement
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Call a POISON CENTER or doctor/physician if you feel u
EINECSNumber 203-545-4

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit