Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Vinyl benzoate, 95%

Click to view available options
Quantity:
100g
500g
769-78-8
C9H8O2
148.161
MFCD00048141
KOZCZZVUFDCZGG-UHFFFAOYSA-N
vinyl benzoate,benzoic acid vinyl ester,benzoic acid, ethenyl ester,benzoic acid, vinyl ester,unii-f1e7c1ggku,vinylester kyseliny benzoove,vinylester kyseliny benzoove czech,f1e7c1ggku,benzoic acid, ethenyl ester 9ci,benzoic acid vinyl
13037
CHEBI:84279
ethenyl benzoate
C=COC(=O)C1=CC=CC=C1
769-78-8
C9H8O2
148.161
MFCD00048141
KOZCZZVUFDCZGG-UHFFFAOYSA-N
vinyl benzoate,benzoic acid vinyl ester,benzoic acid, ethenyl ester,benzoic acid, vinyl ester,unii-f1e7c1ggku,vinylester kyseliny benzoove,vinylester kyseliny benzoove czech,f1e7c1ggku,benzoic acid, ethenyl ester 9ci,benzoic acid vinyl
13037
CHEBI:84279
ethenyl benzoate
C=COC(=O)C1=CC=CC=C1
Applications
It is used as chemical reagent and pharmaceutical intermediates.

Solubility
Soluble in methanol

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Quantity 500g
Boiling Point 73°C to 74°C (8mmHg)
Flash Point 82°C (180°F)
Beilstein 2041125
Formula Weight 148.16
Percent Purity 95%
Chemical Name or Material Vinyl benzoate

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DI1050000