Learn More
Fisherbrand™ Vinyl Chair Chrome - Desk Height with Adjustable Arms and Casters in Grade B Vinyl Products for Science Education
Fisherbrand™ Vinyl Chairs in Chrome are height adjustable for 31 in. work surfaces & feature ergonomic adjustments, adjustable arms, & self-braking casters. Antibacterial/Antifungal Grade B vinyl in a variety of colors will suit any work space
$353.00
Specifications
Adjustable Yes (Pneumatic)
Material Vinyl
Footrest No
With Armrest Yes
With Backrest Yes
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S36636 Products for Science Education
View Documents
Blue
Each for $353.00
Add to cart
S36921 Products for Science Education
View Documents
Black
Each for $353.00
Add to cart
S36390 Products for Science Education
View Documents
Burgundy
Each for $353.00
Add to cart
S37039 Products for Science Education
View Documents
Gray
Each for $353.00
Add to cart
S36391 Products for Science Education
View Documents
Dark Blue
Each for $353.00
Add to cart
Description
Description
  • Adjustable from 19-24 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
Specifications
Specifications
Yes (Pneumatic)
No
Yes
No
Vinyl
Yes
Yes