Learn More
Fisherbrand™ Vinyl Chair Chrome - Desk Height with Seat Tilt and Casters in Grade B Vinyl Products for Science Education
Fisherbrand™ Vinyl Chairs in Chrome are height adjustable for 31 in. work surfaces & feature ergonomic adjustments & self-braking casters. Antibacterial/Antifungal Grade B vinyl in a variety of colors will suit any work space
 View more versions of this product
Catalog No. S36444
Add to cart
Description
Description
  • Adjustable from 19-24 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Locking seat tilt
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
Specifications
Specifications
Standard
Polished Cast Aluminum
136 kg
18 in.
No
General Seating Chair
Desk Height
15 year parts, lifetime on pneumatic lift, 5 year on upholstery
19 in.
No
Yes
19 to 24 in.
Yes (Pneumatic)
300 lb.
Blue
Chrome-hooded dual-wheel, self-braking, non-marring casters (suitable for both hard and carpeted floors)
Trailblazer Grade B Vinyl
Ergonomic 8 degree forward and 3 degree backward seat tilt locks in place
2.5 in.
32 lb.
25 in.
Yes
Yes