Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Fisherbrand™ Vinyl Chair Chrome - Medium Bench Height with Adjustable Arms, Chrome Foot Ring, and Casters in Grade B Vinyl Products for Science Education
Fisherbrand™ Vinyl Chairs in Chrome are height adjustable for 36 in. work surfaces & feature ergonomic adjustments, adjustable arms, chrome adjustable foot ring, & self-braking casters. Antibacterial/Antifungal Grade B vinyl in a variety of colors will suit any work space.
Specifications
Adjustable Yes (Pneumatic)
Material Vinyl
Footrest Yes
With Armrest Yes
With Backrest Yes
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S36437 Products for Science Education
View Documents
Blue
Each for $403.00
N/A
S36446 Products for Science Education
View Documents
Black
Each for $403.00
N/A
S38485 Products for Science Education
View Documents
Burgundy
Each for $403.00
N/A
S36439 Products for Science Education
View Documents
Gray
Each for $403.00
N/A
S38486 Products for Science Education
View Documents
Dark Blue
Each for $403.00
N/A
Description
Description
  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Chrome adjustable foot ring
  • Cast-aluminum base
  • Chrome-hooded, dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
Specifications
Specifications
Yes (Pneumatic)
Yes
Yes
No
Vinyl
Yes
Yes