Learn More
Fisherbrand™ Vinyl Chair - Desk Height with Casters in Grade B Vinyl Products for Science Education
Fisherbrand™ Vinyl Chairs are height adjustable for 31 in. work surfaces and feature ergonomic adjustments and self-braking casters. Antibacterial/Antifungal Grade B vinyl in a variety of colors will suit any work space
$234.00
Specifications
Adjustable Yes (Pneumatic)
Base Material Nylon Reinforced Fiberglass
Material Vinyl
Footrest No
With Armrest No
View More Specs
Products 4
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S0100474 Products for Science Education
View Documents
Taupe
Each for $234.00
Add to cart
S0100491 Products for Science Education
View Documents
Dove
Each for $234.00
Add to cart
S0100511 Products for Science Education
View Documents
Orange
Each for $234.00
Add to cart
S0100717 Products for Science Education
View Documents
White
Each for $234.00
Add to cart
Description
Description
  • Adjustable from 18-23 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Black, nylon-reinforced fiberglass base
  • Dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
Specifications
Specifications
Yes (Pneumatic)
Vinyl
No
Yes
Desk Height
Nylon Reinforced Fiberglass
No
Yes
No