Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Vinyl Chair - Desk Height with Round Tube Base, Seat Tilt
SDPSCI_ED

Fisherbrand™ Vinyl Chairs with Round Tube Base are height adjustable for 31 in. work surfaces and feature ergonomic adjustments

 View more versions of this product

Catalog No. S0100539


Add to Cart

Description

Description

  • Adjustable from 20.5 to 25.75 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward 8-degree forward and 3-degree backward locking seat tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Black, powder-coated steel base with fixed foot ring
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade A Vinyl
  • Additional antibacterial protections
  • High end look and feel
  • Variety of color options to suit any work space
Specifications

Specifications

General Purpose Seating
Powder-coated Steel
6-way adjustable up and down, in and out, twist toward or away from the user
Desk Height
18 in.
Ergonomic 8 degree forward and 3 degree backward seat tilt locks in place
2.5 in.
Yes
No
42 lb.
Yes
Yes
20.5 to 25.75 in.
Yes (Pneumatic)
Taupe
Powder-coated steel with fixed foot ring
Non-marring bell glides (suitable for both hard and carpeted floors)
Round tube chrome foot ring with knob lock
25 in.
300 lb.
19 in.
15 year parts, lifetime on pneumatic lift, 5 year on upholstery
No
Vinyl
Recommendations

Recommendations

Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit