Learn More
Fisherbrand™ Vinyl Chair - Med Bench Height with Medium Back, Seat Tilt, Adj Arms, Chrome Foot Ring, and Stationary Glides in Grade B Vinyl Products for Science Education
Fisherbrand™ Vinyl Chairs with Medium Back are height adjustable for 36 in.work surfaces and feature ergonomic adjustments, larger back, adjustable arms, chrome adjustable foot ring, and stationary glides. Antibacterial/Antifungal Grade B vinyl in a variety of colors will suit any work space.
$421.00
Specifications
Adjustable Yes (Pneumatic)
Base Style Nylon Reinforced Fiberglass
Material Trailblazer Grade B Vinyl
Footrest Yes
With Armrest Yes
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S38480 Products for Science Education
View Documents
Blue
Each for $421.00
Add to cart
S36235 Products for Science Education
View Documents
Black
Each for $421.00
Add to cart
S36243 Products for Science Education
View Documents
Burgundy
Each for $421.00
Add to cart
S36693 Products for Science Education
View Documents
Gray
Each for $421.00
Add to cart
S36244 Products for Science Education
View Documents
Dark Blue
Each for $421.00
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 21-28.5 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward 8-degree forward and 3-degree backward locking seat tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Black, nylon-reinforced fiberglass base
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
Specifications
Specifications
Yes (Pneumatic)
Trailblazer Grade B Vinyl
Yes
No
Medium Bench Height
Nylon Reinforced Fiberglass
Yes
Yes
Yes