Learn More
Fisherbrand™ Vinyl Chair - Medium Bench Height with Medium Back, Adjustable Arms, Chrome Foot Ring, and Casters in Grade B Vinyl Products for Science Education
Fisherbrand™ Vinyl Chairs with Medium Back are height adjustable for 36 in.work surfaces & feature ergonomic adjustments, larger back, adjustable arms, chrome adjustable foot ring, & self-braking casters. Antibacterial/Antifungal Grade B vinyl in a variety of colors will suit any work space.
$400.00
Specifications
Adjustable Yes (Pneumatic)
Material Vinyl
Footrest Yes
With Armrest Yes
With Backrest Yes
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S38509 Products for Science Education
View Documents
Blue
Each for $400.00
Add to cart
S38637 Products for Science Education
View Documents
Black
Each for $400.00
Add to cart
S38638 Products for Science Education
View Documents
Burgundy
Each for $400.00
Add to cart
S38636 Products for Science Education
View Documents
Gray
Each for $400.00
Add to cart
S38462 Products for Science Education
View Documents
Dark Blue
Each for $400.00
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 21-28.5 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Chrome adjustable foot ring
  • Black, nylon-reinforced fiberglass base
  • Dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
Specifications
Specifications
Yes (Pneumatic)
Yes
Yes
No
Vinyl
Yes
Yes