Learn More
Fisherbrand™ Vinyl Chair - Medium Bench Height with Medium Back, Chrome Foot Ring, and Casters in Grade B Vinyl Products for Science Education
Fisherbrand™ Vinyl Chairs with Medium Back are height adjustable for 36 in.work surfaces and feature ergonomic adjustments, larger back, chrome adjustable foot ring, and self-braking casters. Antibacterial/Antifungal Grade B vinyl in a variety of colors will suit any work space.
$319.00
Specifications
Adjustable Yes (Pneumatic)
Base Material Nylon Reinforced Fiberglass
Material Vinyl
Footrest Yes
With Armrest No
View More Specs
Products 4
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S0101262 Products for Science Education
View Documents
White
Each for $319.00
Add to cart
S0101286 Products for Science Education
View Documents
Dove
Each for $319.00
Add to cart
S0101310 Products for Science Education
View Documents
Taupe
Each for $319.00
Add to cart
S0101334 Products for Science Education
View Documents
Orange
Each for $319.00
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 21-28.5 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Black, nylon-reinforced fiberglass base
  • Dual-wheel, self-braking, non-marring casters suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
Specifications
Specifications
Yes (Pneumatic)
Vinyl
No
Yes
Medium Bench Height
Nylon Reinforced Fiberglass
Yes
Yes
No