Learn More
Fisherbrand™ Vinyl Chair - Medium Bench Height with Round Tube Base, Adjustable Arms, and Stationary Glides in Grade B Vinyl Products for Science Education
Fisherbrand™ Vinyl Chairs with Round Tube Base are height adjustable for 36 in. work surfaces and feature ergonomic adjustments, adjustable arms, fixed foot ring, and stationary glides. Antibacterial/Antifungal Grade B vinyl in a variety of colors will suit any work space.
$366.00
Specifications
Adjustable Yes (Pneumatic)
Material Vinyl
Footrest No
With Armrest Yes
With Backrest Yes
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S36371 Products for Science Education
View Documents
Black
Each for $366.00
Add to cart
S36468 Products for Science Education
View Documents
Black
Each for $366.00
Add to cart
S36470 Products for Science Education
View Documents
Black
Each for $366.00
Add to cart
S36372 Products for Science Education
View Documents
Black
Each for $366.00
Add to cart
S36477 Products for Science Education
View Documents
Black
Each for $366.00
Add to cart
Description
Description
  • Adjustable from 23.25-31 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Black, powder-coated steel base with fixed foot ring
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
Specifications
Specifications
Yes (Pneumatic)
No
Yes
No
Vinyl
Yes
No