Learn More
Fisherbrand™ Vinyl Chair - Medium Bench Height with Round Tube Base, Medium Back, and Stationary Glides in Grade B Vinyl Products for Science Education
Fisherbrand™ Vinyl Chairs with Medium Back and Round Tube Base are height adjustable for 36 in. work surfaces and feature ergonomic adjustments, larger back, fixed foot ring, and stationary glides. Antibacterial/Antifungal Grade B vinyl in a variety of colors will suit any work space.
$306.00
Specifications
Adjustable Yes (Pneumatic)
Base Material Powder-coated Steel
Material Vinyl
Footrest No
With Armrest No
View More Specs
Products 4
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S0101268 Products for Science Education
View Documents
Black
Each for $306.00
Add to cart
S0101292 Products for Science Education
View Documents
Black
Each for $306.00
Add to cart
S0101316 Products for Science Education
View Documents
Black
Each for $306.00
Add to cart
S0101340 Products for Science Education
View Documents
Orange
Each for $306.00
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 23.25-31 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Black, powder-coated steel base with fixed foot ring
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
Specifications
Specifications
Yes (Pneumatic)
Vinyl
No
No
Medium Bench Height
Powder-coated Steel
No
Yes
No