Learn More
Vinyl-Coated Lead Rings Products for Science Education
Stabilize flasks in baths and on the bench.
$49.00
Specifications
Autoclavable Not Autoclavable
Type Accessory
View More Specs
Products 1
Catalog Number Mfr. No. Diameter (English) Inner Diameter (Metric) Inner Weight (English) Weight (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (English) Inner Diameter (Metric) Inner Weight (English) Weight (Metric) Price Quantity & Availability  
S90140B Products for Science Education
View Documents
Bel-Art - SP Scienceware
F183081000/EMD
2 in. 5.08 cm 1 lb. 450 g
Each for $49.00
Add to cart
Description
Description
  • Chemical-resistant, vinyl coating prevents glass damage and slippage
  • Multiple sizes and weights available for flasks up to 1.05 gal. (4L)
  • Position on sidearm flasks without disconnecting tubing
  • Adjust diameter to best fit flask or other vessel
  • Use to reduce breakage of glass cylinders by placing around base
Specifications
Specifications
Not Autoclavable
Accessory