Learn More
Fisher Science Education™ Vinyl Desk-Height Chair with Round Tube Base, Ajustable Arms and Glides Products for Science Education
6-way adjustable arm rests support a variety of arm placements for proper ergonomics. Desk-height chairs are ideal for work surface height range of 29 to 31 in.
$333.00 - $369.00
Specifications
Adjustable (Height) 19 to 24 in. (48.26 to 60.96cm)
Material Vinyl
Adjustable Arms
Seating Height 29 to 31 in.
Back Style Standard
View More Specs
Products 8
Catalog Number Color Seat Tilt Price Quantity & Availability  
Catalog Number Color Seat Tilt Price Quantity & Availability  
S36417 Products for Science Education View Documents Burgundy Yes Each for $369.00
Add to cart
S36538 Products for Science Education View Documents Navy Yes Each for $369.00
Add to cart
S36577 Products for Science Education View Documents Blue Yes Each for $369.00
Add to cart
S36578 Products for Science Education View Documents Gray Yes Each for $369.00
Add to cart
S36579 Products for Science Education View Documents Black Yes Each for $369.00
Add to cart
S36800 Products for Science Education View Documents Gray No Tilt Each for $333.00
Add to cart
S36801 Products for Science Education View Documents Black No Tilt Each for $333.00
Add to cart
S38443 Products for Science Education View Documents Blue No Tilt Each for $333.00
Add to cart
Description
Description
  • Adjustable arm rests move up and down, in and out, and twist toward or away from the user
  • Constructed of highly durable molded polyurethane foam cushioning covered with easily cleaned antibacterial/anti-microbial vinyl
  • Contoured waterfall seat cushion and an adjustable backrest with lumbar support for increased comfort during extended sitting durations
  • 25 in. (63.5cm) 5-leg black powder coated steel base with fixed foot ring
  • Black, non-marring bell glides are suitable for both hard and carpeted floors
  • Optional seat tilt provides 8° forward and 3° backward locking tilt for ergonomic posture support
  • Available in 5 standard colors
Warranty and Services
15-year warranty, lifetime on the pneumatic lift
Specifications
Specifications
19 to 24 in. (48.26 to 60.96cm)
Arms
Standard
19 in.
2.5 in.
12 in.
25 in.
300 lb.
Vinyl
29 to 31 in.
5-leg black powder coated steel base with fixed foot ring
18 in.
17.5 in.
2.5 in.
63.5cm