Learn More
Fisherbrand Vinyl ESD Chair - Desk Height with Adjustable Arms and ESD Casters in ESD Vinyl
Fisherbrand Vinyl ESD Chairs are height adjustable for 31 in. work surfaces & feature ergonomic adjustments, electro-static components such as Vinyl & self-braking casters, & adjustable arms. Chairs are manufactured to ensure proper grounding for static sensitive applications.
$478.00
Specifications
Adjustable | Yes |
---|---|
Base Style | Polished Cast Aluminum |
Material | Polyurethane foam cushioning covered with static-dissipating, heavy duty vinyl |
Footrest | No |
With Armrest | Yes |
Description
- Adjustable from 19-24 inches from the top of the seat to the floor
- Features ergonomic adjustments including seat height
- Locking back tilt
- 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
- Electro-static dissipating components to ensure proper grounding
- Cast-aluminum base
- Dual-wheel, self-braking, non-marring ESD casters suitable for hard floors
- Electro-static dissipating vinyl
- Meets ANSI/ESD STM12.1-2013
Chairs shipping to California or Massachusetts may be subject to state mandated fire treatment and may be subject to an up-charge.
Specifications
Yes | |
Polyurethane foam cushioning covered with static-dissipating, heavy duty vinyl | |
Yes | |
No | |
ANSI/ESD-STM-12.1-2006 |
Polished Cast Aluminum | |
No | |
Yes | |
Desk Height |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.