Learn More
Fisherbrand™ Vinyl ESD Chair - Desk Height with Seat Tilt and ESD Stationary Glides in ESD Vinyl Products for Science Education
Fisherbrand™ Vinyl ESD Chairs are height adjustable for 31 in. work surfaces and feature ergonomic adjustments and electro-static components such as Vinyl and stationary glides. Chairs are manufactured to ensure proper grounding for static sensitive applications.
$419.00
Specifications
Adjustable Yes
Footrest No
With Armrest No
With Casters No
With Tilt Yes
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S36170 Products for Science Education
View Documents
Black
Each for $419.00
Add to cart
S36175 Products for Science Education
View Documents
Blue
Each for $419.00
Add to cart
Description
Description
  • Adjustable from 19-24 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • Electro-static dissipating components to ensure proper grounding
  • Cast-aluminum base
  • Stationary ESD glides suitable for hard floors
  • Electro-static dissipating vinyl
  • Meets ANSI/ESD STM12.1-2013
Chairs shipping to California or Massachusetts may be subject to state mandated fire treatment and may be subject to an up-charge.
Specifications
Specifications
Yes
No
Yes
No
No
ANSI/ESD-STM-12.1-2006