Learn More
Fisherbrand™ Vinyl ESD Chair - Medium Bench Height with Adjustable Arms and ESD Casters in ESD Vinyl Products for Science Education
Fisherbrand™ Vinyl ESD Chairs are height adjustable for 36 in. work surfaces & feature ergonomic adjustments, electro-static components such as Vinyl & self-braking casters, & adjustable arms. Chairs are manufactured to ensure proper grounding for static sensitive applications.
$496.00
Specifications
Adjustable Yes
Footrest No
With Armrest Yes
With Casters Yes
With Tilt No
View More Specs
Products 2
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S36401 Products for Science Education
View Documents
Blue
Each for $496.00
Add to cart
S36400 Products for Science Education
View Documents
Black
Each for $496.00
Add to cart
Description
Description
  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 6-way adjustable arm rests adjust up and down, in and out, and twist toward and away from the user
  • Electro-static dissipating components to ensure proper grounding
  • Electro-static dissipating vinyl
  • Meets ANSI/ESD STM12.1-2013
Chairs shipping to California or Massachusetts may be subject to state mandated fire treatment and may be subject to an up-charge.
Specifications
Specifications
Yes
Yes
No
No
Yes
ANSI/ESD-STM-12.1-2006