Learn More
Fisherbrand Vinyl ESD Chair - Medium Bench Height with Chrome Foot Ring and ESD Stationary Glides in ESD Vinyl
Fisherbrand Vinyl ESD Chairs are height adjustable for 36 in. work surfaces and feature ergonomic adjustments and electro-static components such as Vinyl and stationary glides. Chairs are manufactured to ensure proper grounding for static sensitive applications.
$439.00
Specifications
Adjustable | Yes |
---|---|
Base Style | Polished Cast Aluminum |
Material | Polyurethane foam cushioning covered with static-dissipating, heavy duty vinyl |
Footrest | Yes |
With Armrest | No |
Description
- Adjustable from 21.5-29 inches from the top of the seat to the floor
- Features ergonomic adjustments including seat height
- Locking back tilt
- 20-Inch Chrome adjustable foot ring with internal lock
- Electro-static dissipating components to ensure proper grounding
- Cast-aluminum base
- Stationary ESD glides suitable for hard floors
- Electro-static dissipating vinyl
- Meets ANSI/ESD STM12.1-2013
Chairs shipping to California or Massachusetts may be subject to state mandated fire treatment and may be subject to an up-charge.
Specifications
Yes | |
Polyurethane foam cushioning covered with static-dissipating, heavy duty vinyl | |
No | |
No | |
ANSI/ESD-STM-12.1-2006 |
Polished Cast Aluminum | |
Yes | |
No | |
Medium Bench Height |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.