Learn More
Fisherbrand™ Vinyl ESD Chair - Medium Bench Height with ESD Casters in ESD Vinyl
SCI_ED
Fisherbrand™ Vinyl ESD Chairs are height adjustable for 36 in. work surfaces and feature ergonomic adjustments, electro-static components such as Vinyl and self-braking casters, and adjustable arms. Chairs are manufactured to ensure proper grounding for static sensitive applications.
 View more versions of this product
Catalog No. S38719
Add to cart
Description
Description
  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Electro-static dissipating components to ensure proper grounding
  • Cast-aluminum base
  • Dual-wheel, self-braking, non-marring ESD casters suitable for hard floors
  • Electro-static dissipating vinyl
  • Meets ANSI/ESD STM12.1-2013
Chairs shipping to California or Massachusetts may be subject to state mandated fire treatment and may be subject to an up-charge.
Specifications
Specifications
Standard
ANSI/ESD-STM-12.1-2006
Electro-Static Dissipating, dual-wheel, non-marring casters (suitable for both hard and carpeted floors)
Polyurethane foam, heavy-duty ESD fabric
32 lb.
Yes
21.5 to 29 in.
ESD Chair
Yes
Black
No
Medium Bench Height
No
No
Polished Cast Aluminum