Learn More
Fisherbrand™ Vinyl ESD Chair - Medium Bench Height with Medium Back, Chrome Foot Ring, and ESD Stationary Glides in ESD Vinyl
SCI_ED
Fisherbrand™ Vinyl ESD Chairs with Medium Back are height adjustable for 36 in. work surfaces and feature ergonomic adjustments and electro-static components such as Vinyl and stationary glides. Chairs with Medium Back are manufactured to ensure proper grounding for static sensitive applications.
 View more versions of this product
Catalog No. S39928
Add to cart
Description
Description
  • Medium Back
  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Electro-static dissipating components to ensure proper grounding
  • Cast-aluminum base
  • Stationary ESD glides suitable for hard floors
  • Electro-static dissipating vinyl
  • Meets ANSI/ESD STM12.1-2013
Specifications
Specifications
Standard
ANSI/ESD-STM-12.1-2006
Electro-Static Dissipating, non-marring bell glides (suitable for both hard and carpeted floors)
20 in.
Polyurethane foam, heavy-duty ESD fabric
35 lb.
Yes
No
300 lb.
ESD Chair
Yes
Black
Yes
Round tube chrome foot ring with internal lock
Medium Bench Height
No
No
21.5 to 29 in.
Polished Cast Aluminum
136 kg