Learn More
Fisherbrand™ Vinyl ESD Chair - Medium Bench Height with Seat Tilt, Chrome Foot Ring, and ESD Stationary Glides in ESD Vinyl
SCI_ED
Fisherbrand™ Vinyl ESD Chairs are height adjustable for 36 in. work surfaces and feature ergonomic adjustments and electro-static components such as Vinyl and stationary glides. Chairs are manufactured to ensure proper grounding for static sensitive applications.
 View more versions of this product
Catalog No. S36945
Add to cart
Description
Description
  • Adjustable from 21.5-29 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • 20-Inch Chrome adjustable foot ring with internal lock
  • Electro-static dissipating components to ensure proper grounding
  • Cast-aluminum base
  • Stationary ESD glides suitable for hard floors
  • Electro-static dissipating vinyl
  • Meets ANSI/ESD STM12.1-2013
Chairs shipping to California or Massachusetts may be subject to state mandated fire treatment and may be subject to an up-charge.
Specifications
Specifications
Standard
ANSI/ESD-STM-12.1-2006
Electro-Static Dissipating, non-marring bell glides (suitable for both hard and carpeted floors)
Round tube chrome foot ring with internal lock
Polyurethane foam, heavy-duty ESD fabric
35 lb.
No
21.5 to 29 in.
ESD Chair
Yes
Black
Yes
Ergonomic 8 degree forward and 3 degree backward seat tilt locks in place
Medium Bench Height
No
Yes
Polished Cast Aluminum