Learn More
Fisher Science Education™ Vinyl ESD Chrome Chairs with Adjustable Arms, Desk Height Products for Science Education
Vinyl-upholstered chair offers ESD protections.
$478.00 - $509.00
Products 7
Catalog Number With Tilt Foot Type Color Price Quantity & Availability  
Catalog Number With Tilt Foot Type Color Price Quantity & Availability  
S36467 Products for Science Education View Documents No tilt No Glides Black Each for $478.00
Add to cart
S36516 Products for Science Education View Documents No tilt ESD Glides Blue Each for $478.00
Add to cart
S36521 Products for Science Education View Documents Standard tilt ESD glides Black Each for $509.00
Add to cart
S36753 Products for Science Education View Documents Standard Tilt ESD Glides Blue Each for $509.00
Add to cart
S38473 Products for Science Education View Documents Standard Tilt No glides Black Each for $509.00
Add to cart
S38474 Products for Science Education View Documents Standard tilt No glides Blue Each for $509.00
Add to cart
S38589 Products for Science Education View Documents No tilt No glides Blue Each for $478.00
Add to cart
Description
Description
  • Ergonomic
  • Features proper grounding, conductive, and dissipative static control materials
  • Additional ergonomic benefits include contoured waterfall cushion and adjustable back rest with lumbar support for increased comfort during extended sitting durations
  • Optional seat tilt provides 8° forward and 3° backward locking tilt for ergonomic posture support
  • Foot rings are available in 20 in. diameter chrome
  • Desk height ESD chairs feature several customizable options, including choice of ESD casters or glides on polished cast aluminum base, and static-dissipating color configurations
Warranty and Services
15-year warranty on chair, lifetime on pneumatic cylinder, 5 year upholstery
Chairs shipping to California or Massachusetts may be subject to state mandated fire treatment and may be subject to an up-charge.