Learn More
Fisher Science Education™ Vinyl ESD Chrome Chairs, Desk Height Products for Science Education
Vinyl-upholstered chair offers ESD protections.
$389.00 - $419.00
Products 7
Catalog Number Color With Tilt Foot Type Price Quantity & Availability  
Catalog Number Color With Tilt Foot Type Price Quantity & Availability  
S36191 Products for Science Education View Documents Each for $419.00
Add to cart
S36211 Products for Science Education View Documents Each for $389.00
Add to cart
S36566 Products for Science Education View Documents Each for $419.00
Add to cart
S36170 Products for Science Education View Documents Each for $419.00
Add to cart
S38733 Products for Science Education View Documents Each for $389.00
Add to cart
S36175 Products for Science Education View Documents Each for $419.00
Add to cart
S38735 Products for Science Education View Documents Each for $389.00
Add to cart
Description
Description
  • Ergonomic
  • Features proper grounding, conductive, and dissipative static control materials
  • Additional ergonomic benefits include contoured waterfall cushion and adjustable back rest with lumbar support for increased comfort during extended sitting durations
  • Optional seat tilt provides 8° forward and 3° backward locking tilt for ergonomic posture support
  • Foot rings are available in 20 in. diameter chrome
  • Desk height ESD chairs feature several customizable options, including choice of ESD casters or glides on polished cast aluminum base, and static-dissipating color configurations
Warranty and Services
15-year warranty on chair, lifetime on pneumatic cylinder, 5 year upholstery
Chairs shipping to California or Massachusetts may be subject to state mandated fire treatment and may be subject to an up-charge.