Learn More
Fisher Science Education™ Vinyl ESD Chrome Chairs with Foot Ring, High Bench Height Products for Science Education
Vinyl-upholstered chair offers ESD protections.
$469.00 - $500.00
Products 8
Catalog Number Color With Tilt Foot Type Price Quantity & Availability  
Catalog Number Color With Tilt Foot Type Price Quantity & Availability  
S36033 Products for Science Education View Documents Each for $500.00
Add to cart
S36571 Products for Science Education View Documents Each for $469.00
Add to cart
S36666 Products for Science Education View Documents Each for $500.00
Add to cart
S36572 Products for Science Education View Documents Each for $469.00
Add to cart
S36780 Products for Science Education View Documents Each for $500.00
Add to cart
S36948 Products for Science Education View Documents Each for $469.00
Add to cart
S38452 Products for Science Education View Documents Each for $469.00
Add to cart
S38659 Products for Science Education View Documents Each for $500.00
Add to cart
Description
Description
  • Ergonomic
  • Features proper grounding, conductive, and dissipative static control materials
  • Additional ergonomic benefits include contoured waterfall cushion and adjustable back rest with lumbar support for increased comfort during extended sitting durations
  • Optional seat tilt provides 8° forward and 3° backward locking tilt for ergonomic posture support
  • Foot rings are available in 20 in. diameter chrome
  • High bench height ESD chairs feature several customizable options, including choice of ESD casters or glides on polished cast aluminum base, and static-dissipating color configurations
Warranty and Services
15-year warranty on chair, lifetime on pneumatic cylinder, 5 year upholstery
Chairs shipping to California or Massachusetts may be subject to state mandated fire treatment and may be subject to an up-charge.