Learn More
Fisher Science Education™ Vinyl Medium Bench-Height Chair with Chrome Base and Glides, Medium Back Products for Science Education
Armless medium bench-height chairs are ideal for work surface height range of 32 to 36 in. Medium back offers a larger back height of 15 in.
$328.00 - $359.00
Specifications
Adjustable (Height) 22 to 29 in. (55.88 to 73.66cm)
Material Vinyl
Product Type Pneumatic Chair
Seating Height 32 to 36 in.
Back Style Medium
View More Specs
Products 9
Catalog Number Color Seat Tilt Price Quantity & Availability  
Catalog Number Color Seat Tilt Price Quantity & Availability  
S36087 Products for Science Education View Documents Each for $328.00
Add to cart
S36133 Products for Science Education View Documents Each for $359.00
Add to cart
S36224 Products for Science Education View Documents Gray No Tilt Each for $328.00
Add to cart
S36723 Products for Science Education View Documents Navy No Tilt Each for $328.00
Add to cart
S37025 Products for Science Education View Documents Each for $328.00
Add to cart
S37026 Products for Science Education View Documents Burgundy No Tilt Each for $328.00
Add to cart
S38687 Products for Science Education View Documents Each for $359.00
Add to cart
S38688 Products for Science Education View Documents Each for $359.00
Add to cart
S38689 Products for Science Education View Documents Each for $359.00
Add to cart
Description
Description
  • Constructed of highly durable molded polyurethane foam cushioning covered with easily cleaned antibacterial/anti-microbial vinyl
  • Contoured waterfall seat cushion and an adjustable backrest with lumbar support for increased comfort during extended sitting durations
  • 26 in. (66.04cm) 5-leg polished cast aluminum
  • Black, non-marring bell glides are suitable for both hard and carpeted floors
  • Optional seat tilt provides 8° forward and 3° backward locking tilt for ergonomic posture support
  • Available in 5 standard colors
Warranty and Services
15-year warranty, lifetime on the pneumatic lift
Specifications
Specifications
22 to 29 in. (55.88 to 73.66cm)
Pneumatic Chair
Medium
19 in.
2.5 in.
15 in.
26 in.
300 lb.
Vinyl
32 to 36 in.
5-leg polished cast aluminum
18 in.
17.5 in.
2.5 in.
66.04cm