Learn More
Fisher Science Education™ Vinyl Medium Bench-Height Chair with Round Tube Base, Adjustable Arms and Casters, Medium Back Products for Science Education
6-way adjustable arm rests support a variety of arm placements for proper ergonomics. Medium back offers a larger back height of 15 in.
$397.00 - $422.00
Products 8
Catalog Number Color Seat Tilt Price Quantity & Availability  
Catalog Number Color Seat Tilt Price Quantity & Availability  
S36048 Products for Science Education View Documents Blue No Tilt Each for $397.00
Add to cart
S36612 Products for Science Education View Documents Each for $397.00
Add to cart
S36980 Products for Science Education View Documents Each for $397.00
Add to cart
S37027 Products for Science Education View Documents Each for $422.00
Add to cart
S38706 Products for Science Education View Documents Each for $422.00
Add to cart
S36307 Products for Science Education View Documents Each for $397.00
Add to cart
S36164 Products for Science Education View Documents Each for $422.00
Add to cart
S36176 Products for Science Education View Documents Each for $422.00
Add to cart
Description
Description
  • Adjustable arm rests move up and down, in and out, and twist toward or away from the user
  • Constructed of highly durable molded polyurethane foam cushioning covered with easily cleaned antibacterial/anti-microbial vinyl
  • Contoured waterfall seat cushion and an adjustable backrest with lumbar support for increased comfort during extended sitting durations
  • 25 in. (63.5cm) 5-leg black powder coated steel base with fixed foot ring
  • Casters provide mobility on hard surface floors, such as concrete, tile or vinyl
  • Optional seat tilt provides 8° forward and 3° backward locking tilt for ergonomic posture support
  • Available in 5 standard colors
Warranty and Services
15-year warranty, lifetime on the pneumatic lift