Learn More
Fisher Science Education™ Vinyl Medium Bench-Height Chair with Round Tube Base and Glides, Medium Back Products for Science Education
Armless medium bench-height chairs are ideal for work surface height range of 32 to 36 in. Medium back offers a larger back height of 15 in.
$302.00 - $335.00
Products 10
Catalog Number Color Seat Tilt Price Quantity & Availability  
Catalog Number Color Seat Tilt Price Quantity & Availability  
S36247 Products for Science Education View Documents Each for $335.00
Add to cart
S36359 Products for Science Education View Documents Each for $302.00
Add to cart
S36495 Products for Science Education View Documents Each for $335.00
Add to cart
S36496 Products for Science Education View Documents Each for $335.00
Add to cart
S36497 Products for Science Education View Documents Each for $335.00
Add to cart
S36611 Products for Science Education View Documents Each for $335.00
Add to cart
S38683 Products for Science Education View Documents Each for $302.00
Add to cart
S38684 Products for Science Education View Documents Each for $302.00
Add to cart
S38685 Products for Science Education View Documents Each for $302.00
Add to cart
S36357 Products for Science Education View Documents Each for $302.00
Add to cart
Description
Description
  • Constructed of highly durable molded polyurethane foam cushioning covered with easily cleaned antibacterial/anti-microbial vinyl
  • Contoured waterfall seat cushion and an adjustable backrest with lumbar support for increased comfort during extended sitting durations
  • 25 in. (63.5cm) 5-leg black powder coated steel base with fixed foot ring
  • Black, non-marring bell glides are suitable for both hard and carpeted floors
  • Optional seat tilt provides 8° forward and 3° backward locking tilt for ergonomic posture support
  • Available in 5 standard colors
Warranty and Services
15-year warranty, lifetime on the pneumatic lift