Learn More
Fisherbrand™ Vinyl Stool with Back - Desk Height with Round Tube Base, Seat Tilt, and Stationary Glides in Grade A Vinyl Products for Science Education
Fisherbrand™ Vinyl Stools with Back and Round Tube Base are height adjustable for 31 in. work surface and feature ergonomic adjustments, fixed foot ring, and stationary glides. Grade A vinyl, in a variety of colors, offer additional antibacterial properties and a high-end look and feel.
$293.00
Specifications
Adjustable Yes (Pneumatic)
Adjustable (Height) 21 to 26.25 in.
Base Material Powder-coated Steel
Load Bearing Capacity (English) 300 lb.
Foot Type Non-marring bell glides (suitable for both hard and carpeted floors)
View More Specs
Products 6
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S0100439 Products for Science Education
View Documents
Black
Each for $293.00
Add to cart
S0100650 Products for Science Education
View Documents
Black
Each for $293.00
Add to cart
S0100662 Products for Science Education
View Documents
Black
Each for $293.00
Add to cart
S0100667 Products for Science Education
View Documents
Black
Each for $293.00
Add to cart
S0100668 Products for Science Education
View Documents
Black
Each for $293.00
Add to cart
S0100674 Products for Science Education
View Documents
Black
Each for $293.00
Add to cart
Description
Description
  • Adjustable from 23 to 28.25 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • 8-degree forward and 3-degree backward locking seat tilt
  • Black, powder-coated base with fixed foot ring
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade A Vinyl
  • Additional antibacterial protections
  • High end look and feel
  • Variety of color options to suit any work space
Specifications
Specifications
Yes (Pneumatic)
Powder-coated Steel
Non-marring bell glides (suitable for both hard and carpeted floors)
Round tube chrome foot ring with internal lock
Ergonomic 8 degree forward and 3 degree backward seat tilt locks in place
3.5 in.
32 lb.
17 in.
No
21 to 26.25 in.
300 lb.
No
Vinyl
Desk Height
15 year parts, lifetime on pneumatic lift, 5 year on upholstery
25 in.
No
Yes