Learn More
Fisherbrand™ Vinyl Stool with Back - Medium Bench Height with Round Tube Base and Stationary Glides in Grade B Vinyl Products for Science Education
Fisherbrand™ Vinyl Stools with Back and Round Tube Base are height adjustable for 36 in. work surface and feature ergonomic adjustments, fixed foot ring, and stationary glides. Antibacterial/Antimicrobial Grade B vinyl in a variety of colors will suit any work space.
$287.00
Specifications
Adjustable Yes (Pneumatic)
Material Vinyl
Footrest No
With Armrest No
With Backrest Yes
View More Specs
Products 5
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S38508 Products for Science Education
View Documents
Black
Each for $287.00
Add to cart
S36128 Products for Science Education
View Documents
Black
Each for $287.00
Add to cart
S36116 Products for Science Education
View Documents
Black
Each for $287.00
Add to cart
S38694 Products for Science Education
View Documents
Black
Each for $287.00
Add to cart
S38700 Products for Science Education
View Documents
Black
Each for $287.00
Add to cart
Description
Description
  • Adjustable from 23.5-31 inches from the top of the seat to the floor
  • Features ergonomic adjustments including seat height
  • Locking back tilt
  • Black, powder-coated base with fixed foot ring
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
Specifications
Specifications
Yes (Pneumatic)
No
Yes
No
Vinyl
No
No
Stool with Back