Learn More
Fisherbrand™ Vinyl Stool without Back - Desk Height with Round Tube Base and Stationary Glides in Grade B Vinyl Products for Science Education
Fisherbrand™ Vinyl Stools without Back with Round-Tube Base are height adjustable for 31 in. work surfaces and feature 360-degree control mechanism, fixed foot ring, and stationary glides. Antibacterial/Antifungal Grade B vinyl in a variety of colors will suit any work space.
$199.00
Specifications
Adjustable Yes
Base Material Powder-coated Steel
Material Vinyl
Footrest No
With Armrest No
View More Specs
Products 4
Catalog Number Color Price Quantity & Availability  
Catalog Number Color Price Quantity & Availability  
S0100766 Products for Science Education
View Documents
Black
Each for $199.00
Add to cart
S0100776 Products for Science Education
View Documents
Orange
Each for $199.00
Add to cart
S0100779 Products for Science Education
View Documents
Black
Each for $199.00
Add to cart
S0100792 Products for Science Education
View Documents
Black
Each for $199.00
Add to cart
Description
Description
  • Adjustable from 19.75-25.25 inches from the top of the seat to the floor
  • Features ergonomic adjustments, larger back, including 360-degree, all-around control
  • Black, powder-coated steel base with fixed foot ring
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
Specifications
Specifications
Yes
Vinyl
No
No
Stool
Powder-coated Steel
No
No
No