Learn More
Fisherbrand™ Vinyl Stool without Back - Medium Bench Height with Round Tube Base and Stationary Glides in Grade B Vinyl
SCI_ED
Fisherbrand™ Vinyl Stools without Back with Round-Tube Base are height adjustable for 36 in. work surfaces and feature 360-degree control mechanism, fixed foot ring, and stationary glides. Antibacterial/Antifungal Grade B vinyl in a variety of colors will suit any work space.
 View more versions of this product
Catalog No. S0100732
Add to cart
Description
Description
  • Adjustable from 22.5-30.25 inches from the top of the seat to the floor
  • Features ergonomic adjustments, larger back, including 360-degree, all-around control
  • Black, powder-coated steel base with fixed foot ring
  • Stationary, non-marring glides suitable for hard floors
  • Antibacterial/antifungal Grade B Vinyl
  • Variety of color options to suit any work space
Specifications
Specifications
Yes
Black
No
Stool
No
No
300 lb.
General Purpose Seating
Powder-coated steel with fixed foot ring
Non-marring bell glides (suitable for both hard and carpeted floors)
Vinyl
No
No
22.5 to 30.25 in.
Powder-coated Steel
136 kg