Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyl Trifluoroacetate (stabilized with TBC) 97.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
433-28-3
C4H3F3O2
140.061
MFCD00003561
ZBGRMWIREQJHPK-UHFFFAOYSA-N
vinyl trifluoroacetate,vinyltrifluoroacetate,trifluoroacetic acid vinyl ester,pubchem13129,acmc-1ahnj,vinyl 2,2,2-trifluoroacetate,zbgrmwireqjhpk-uhfffaoysa
79011
ethenyl 2,2,2-trifluoroacetate
C=COC(=O)C(F)(F)F
433-28-3
C4H3F3O2
140.061
MFCD00003561
ZBGRMWIREQJHPK-UHFFFAOYSA-N
vinyl trifluoroacetate,vinyltrifluoroacetate,trifluoroacetic acid vinyl ester,pubchem13129,acmc-1ahnj,vinyl 2,2,2-trifluoroacetate,zbgrmwireqjhpk-uhfffaoysa
79011
ethenyl 2,2,2-trifluoroacetate
C=COC(=O)C(F)(F)F
Quantity 25g
Boiling Point 42°C
Color Yellow
UN Number 2924
Formula Weight 140.06
Physical Form Clear Liquid at 20°C
Percent Purity ≥97.0% (GC)
Chemical Name or Material Vinyl Trifluoroacetate (stabilized with TBC)

TSCA : No

Recommended Storage : Refrigerator