Alfa Aesar™ Vinylboronic acid 2-methyl-2,4-pentanediol ester, 95%, stab. with phenothiazine

Manufacturer: Alfa Aesar™ H5515006

Quantity 5g

 View more versions of this product

Catalog No. AAH5515006

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 4627-10-5
    Formula Weight 154.02
    MDL Number MFCD00066915
    UN Number UN1993
    Synonym 4,4,6-Trimethyl-2-vinyl-1,3,2-dioxaborinane
    Chemical Name or Material Vinylboronic acid 2-methyl-2,4-pentanediol ester, Stabilized with phenothiazine

    Specifications

    Formula Weight 154.02
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Physical Form Liquid
    MDL Number MFCD00066915
    UN Number UN1993
    Synonym 4,4,6-Trimethyl-2-vinyl-1,3,2-dioxaborinane
    Refractive Index 1.4290
    Density 0.893g/mL
    Color Colorless to yellow
    DOT Information Transport Hazard Class: 3; Packaging Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    CAS 4627-10-5
    Health Hazard 1 Warning
    Quantity 5g
    Health Hazard 2 GHS H Statement
    H226-H317
    Flammable liquid and vapor.
    May cause an allergic skin reaction.
    Chemical Name or Material Vinylboronic acid 2-methyl-2,4-pentanediol ester, Stabilized with phenothiazine
    Assay 95%

    GHS H Statement
    H226-H317
    Flammable liquid and vapor.
    May cause an allergic skin reaction.

    GHS P Statement
    P210-P261-P280-P303+P361+P353-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    Store in a well-ventilated place.
    Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit