Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinylidene chloride, 99.9%, stabilized, ACROS Organics™

Click to view available options
Quantity:
1L
Packaging:
Glass bottle
75-35-4,150-76-5
C2H2Cl2
96.938
MFCD00011653
LGXVIGDEPROXKC-UHFFFAOYSA-N
vinylidene chloride,1,1-dichloroethylene,vinylidene dichloride,1,1-dce,ethene, 1,1-dichloro,vinylidine chloride,ethylene, 1,1-dichloro,as-dichloroethylene,rcra waste number u078,asym-dichloroethylene
6366
CHEBI:34031
1,1-dichloroethene
C=C(Cl)Cl
75-35-4,150-76-5
C2H2Cl2
96.938
MFCD00011653
LGXVIGDEPROXKC-UHFFFAOYSA-N
vinylidene chloride,1,1-dichloroethylene,vinylidene dichloride,1,1-dce,ethene, 1,1-dichloro,vinylidine chloride,ethylene, 1,1-dichloro,as-dichloroethylene,rcra waste number u078,asym-dichloroethylene
6366
CHEBI:34031
1,1-dichloroethene
C=C(Cl)Cl
Boiling Point 31.2°C to 31.7°C
Density 1.2180g/mL
Infrared Spectrum Authentic
Quantity 1L
Melting Point -122.0°C
Color Colorless
Stabilizer 180 to 220ppm MEHQ
Water 0.02% max.
Specific Gravity 1.218
Refractive Index 1.4244 to 1.4264
Packaging Glass bottle
Flash Point −25°C
Assay Percent Range 99.90%
Linear Formula H2C=CCl2
Beilstein 01, 186
Merck Index 15, 10192
Formula Weight 96.94
Physical Form Liquid
Percent Purity 99.9%
Chemical Name or Material Vinylidene chloride, 99.9%
Show More Show Less

GHS H Statement:
Suspected of causing cancer.
Harmful if inhaled.
Extremely flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-864-0

RTECSNumber : KV9275000

TSCA : TSCA