Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinylidene chloride, 99.9%, stabilized, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
75-35-4,150-76-5
C2H2Cl2
96.94
MFCD00011653
LGXVIGDEPROXKC-UHFFFAOYSA-N
vinylidene chloride,1,1-dichloroethylene,vinylidene dichloride,1,1-dce,ethene, 1,1-dichloro,vinylidine chloride,ethylene, 1,1-dichloro,as-dichloroethylene,rcra waste number u078,asym-dichloroethylene
6366
CHEBI:34031
1,1-dichloroethene
C=C(Cl)Cl
75-35-4,150-76-5
C2H2Cl2
96.94
MFCD00011653
LGXVIGDEPROXKC-UHFFFAOYSA-N
vinylidene chloride,1,1-dichloroethylene,vinylidene dichloride,1,1-dce,ethene, 1,1-dichloro,vinylidine chloride,ethylene, 1,1-dichloro,as-dichloroethylene,rcra waste number u078,asym-dichloroethylene
6366
CHEBI:34031
1,1-dichloroethene
C=C(Cl)Cl
Viscosity .377 mPa.s (15°C)
Water 0.02% max.
Specific Gravity 1.218
Stabilizer 180 to 220ppm MEHQ
Refractive Index 1.4244 to 1.4264
Packaging Glass bottle
Flash Point −25°C
Boiling Point 31.2°C to 31.7°C
Melting Point -122.0°C
Infrared Spectrum Authentic
Quantity 2.5L
Density 1.2180g/mL
Color Colorless
Assay Percent Range 99.85% min. (GC)
Linear Formula H2C=CCl2
Beilstein 01, 186
Merck Index 15, 10192
Solubility Information Solubility in water: 2.5g/L (25°C). Other solubilities: soluble in most common organic solvents and,greases
Formula Weight 96.94
Physical Form Liquid
Percent Purity 99.9%
Chemical Name or Material Vinylidene chloride, 99.9%
Show More Show Less

GHS H Statement
Suspected of causing cancer.
Harmful if inhaled.
Extremely flammable liquid and vapor.
Harmful if swallowed.
Causes serious eye irritation.
May cause damage to organs through prolonged or repeated expo

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep

GHS Signal Word: Danger

EINECSNumber : 200-864-0

RTECSNumber : KV9275000

TSCA : TSCA