Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinylmagnesium bromide, 0.7M solution in THF, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
1826-67-1, 109-99-9
C2H3BrMg
131.255
MFCD00000042
RMGJCSHZTFKPNO-UHFFFAOYSA-M
vinylmagnesium bromide, bromo ethenyl magnesium, vinyl magnesium bromide, magnesium, bromoethenyl, vinylmagnesium bromide solution, vinylmagnesium bromide solution, 1.0 m in thf, bromovinylmagnesium, bromo vinyl magnesium, vinyl magnesiumbromide, grignard reagent
74584
magnesium;ethene;bromide
C=[CH-].[Mg+2].[Br-]
1826-67-1, 109-99-9
C2H3BrMg
131.255
MFCD00000042
RMGJCSHZTFKPNO-UHFFFAOYSA-M
vinylmagnesium bromide, bromo ethenyl magnesium, vinyl magnesium bromide, magnesium, bromoethenyl, vinylmagnesium bromide solution, vinylmagnesium bromide solution, 1.0 m in thf, bromovinylmagnesium, bromo vinyl magnesium, vinyl magnesiumbromide, grignard reagent
74584
magnesium;ethene;bromide
C=[CH-].[Mg+2].[Br-]
Additional Information magnesium salts may crystallize below 20°C
Linear Formula H2C=CHMgBr
Concentration 0.7 to 0.9M
Density 0.9800g/mL
Formula Weight 131.25
Physical Form Solution
Packaging AcroSeal™ Glass bottle
Solubility Solubility in water: vigorous reaction.
Specific Gravity 0.98
Color Brown
Quantity 100mL
Flash Point −17°C
Chemical Name or Material Vinylmagnesium bromide, 0.7M Solution in THF
Show More Show Less

GHS P Statement

Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Reacts violently with water.

GHS P Statement
Store in a dry place. Store in a closed container.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger