Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vinylmagnesium bromide, 0.7M solution in THF, AcroSeal™, ACROS Organics™
Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
Specifications
Additional Information | magnesium salts may crystallize below 20°C |
Linear Formula | H2C=CHMgBr |
Concentration | 0.7 to 0.9M |
Density | 0.9800g/mL |
Formula Weight | 131.25 |
Physical Form | Solution |
Packaging | AcroSeal™ Glass Bottle |
Solubility | Solubility in water: vigorous reaction. |
Specific Gravity | 0.98 |
Color | Brown |
Show More |
Safety and Handling
GHS P Statement
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Reacts violently with water.
GHS P Statement
Store in a dry place. Store in a closed container.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Danger