Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinyloxytrimethylsilane, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
6213-94-1
C5H12OSi
116.24
MFCD00054764
HFTNNOZFRQLFQB-UHFFFAOYSA-N
trimethyl vinyloxy silane,vinyloxytrimethylsilane,silane, ethenyloxy trimethyl,tmso-ethene,vinyloxy-trimethylsilane,trimethylsiloxy ethylene,snqhhgciiezjp@,acmc-209mzx,ethenoxy trimethyl silane,trimethylsilyl vinyl ether
80344
ethenoxy(trimethyl)silane
C[Si](C)(C)OC=C
6213-94-1
C5H12OSi
116.24
MFCD00054764
HFTNNOZFRQLFQB-UHFFFAOYSA-N
trimethyl vinyloxy silane,vinyloxytrimethylsilane,silane, ethenyloxy trimethyl,tmso-ethene,vinyloxy-trimethylsilane,trimethylsiloxy ethylene,snqhhgciiezjp@,acmc-209mzx,ethenoxy trimethyl silane,trimethylsilyl vinyl ether
80344
ethenoxy(trimethyl)silane
C[Si](C)(C)OC=C
Boiling Point 74°C to 75°C
Specific Gravity 0.77
Flash Point -10°C
Refractive Index 1.3885 to 1.3905
Density 0.7700g/mL
Packaging Glass bottle
Color Yellow
Infrared Spectrum Authentic
Quantity 1g
Assay Percent Range 97%
Linear Formula CH2CHOSi(CH3)3
Solubility Information Solubility in water: insoluble
Formula Weight 116.24
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material Vinyloxytrimethylsilane, 97%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 228-281-7

TSCA : TSCA