Alfa Aesar™ Violuric acid monohydrate, 97%

Manufacturer: Alfa Aesar™ B2393109

Quantity 10g

 View more versions of this product

Catalog No. AAB2393109

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 26351-19-9
    Molecular Formula C4H3N3O4·H2O
    Formula Weight 175.10 (157.08 Anhydrous)
    MDL Number MFCD00149401
    Beilstein 162342
    Merck Index 14,10000
    Synonym Alloxan-5-oxime monohydrate; 5-Isonitrosobarbituric acid monohydrate
    Chemical Name or Material Violuric acid monohydrate

    Specifications

    Formula Weight 175.10 (157.08 Anhydrous)
    TSCA TSCA
    Molecular Formula C4H3N3O4·H2O
    MDL Number MFCD00149401
    CAS 26351-19-9
    Merck Index 14,10000
    Synonym Alloxan-5-oxime monohydrate; 5-Isonitrosobarbituric acid monohydrate
    Melting Point ≈245°C (dec.)
    Health Hazard 1 Warning
    Quantity 10g
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    RTECS Number BA4225000
    Beilstein 162342
    Chemical Name or Material Violuric acid monohydrate
    Assay 97%

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit