Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VIP Antibody (OTI1E8), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274848IR
Description
VIP Monoclonal specifically detects VIP in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
VIP | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
VIP | |
Full length recombinant protein of human VIP (NP_003372) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI1E8 | |
Western Blot, Immunohistochemistry | |
MGC13587, PHM27, vasoactive intestinal peptide, VIP peptides | |
Mouse | |
Affinity Purified | |
Cancer, Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
7432.0 | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction