Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
VIP Antibody (2), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP105163AF350
Description
VIP Monoclonal specifically detects VIP in Human, Rat, Porcine, Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
VIP | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
VIP | |
Vasoactive intestinal peptide adsorbed onto aluminum hydroxide gel | |
0.1 mL | |
Primary | |
Binds to human, bovine, porcine or rat VIP, but does not bind to PHM or PACAP | |
Store at 4°C in the dark. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
2 | |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
MGC13587, PHM27, vasoactive intestinal peptide, VIP peptides | |
Mouse | |
Protein A or G purified | |
Cancer, Cell Cycle and Replication, GPCR, Neuroscience, Neurotransmission, Signal Transduction | |
7432.0 | |
Human, Rat, Pig, Bovine | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction